missing translation for 'onlineSavingsMsg'
Learn More

Pixantrone dimaleate

Product Code. 16657032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16657032 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16657032 Supplier Thermo Scientific Acros Supplier No. 465522500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144675-97-8 | C25H27N5O10 | 557.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector PIXANTRONEDIMALEATE
CAS 144675-97-8
Color Black to Purple
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C25H27N5O10
Quantity 250 mg
Solubility Information (2 mg/ml aq. soln.) Clear solution
InChI Key SVAGFBGXEWPNJC-LVEZLNDCSA-N
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.NCCNC1=CC=C(NCCN)C2=C1C(=O)C1=CC=NC=C1C2=O
IUPAC Name bis((2E)-but-2-enedioic acid); 6,9-bis[(2-aminoethyl)amino]-5H,10H-benzo[g]isoquinoline-5,10-dione
Molecular Weight (g/mol) 557.52
Formula Weight 557.51
Physical Form Powder
Chemical Name or Material Pixantrone dimaleate
Show More Show Less
compliance-icons
Product Identifier
  • Pixantrone dimaleate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Pixantrone dimaleate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.