missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA C alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99335-50ul
This item is not returnable.
View return policy
Description
PKA C alpha Polyclonal antibody specifically detects PKA C alpha in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| PKA C alpha | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:100-1:400 | |
| cAMP-dependent protein kinase catalytic subunit alpha, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PKA C alpha. | |
| 50 μg | |
| Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5566 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction