missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKMYT1 Monoclonal Antibody (OTI2B4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525029
Additional Details : Weight : 0.01000kg
Description
PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.Specifications
PKMYT1 | |
Monoclonal | |
0.88 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99640 | |
Pkmyt1 | |
Full length human recombinant protein of PKMYT1 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
OTI2B4 | |
Unconjugated | |
Pkmyt1 | |
6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg | |
Mouse | |
Affinity Chromatography | |
RUO | |
9088 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |