missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77266MFV610
This item is not returnable.
View return policy
Description
PKR Polyclonal antibody specifically detects PKR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PKR | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PKR. The immunogen is located within the last 50 amino acids of PKR. Amino Acid Squence: TVWKKSPEKNERHT | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Signal Transduction | |
| 5610 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction