Learn More
PLA2G3 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811652S
Description
PLA2G3 Monoclonal antibody specifically detects PLA2G3 in Human samples. It is validated for Western BlotSpecifications
| PLA2G3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H0H5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29127 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4F2 | |
| Unconjugated | |
| RACGAP1 | |
| giii; GIII sPLA2; GIII-SPLA2; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2 group III; phospholipase A2, group III; PLA2G3; SPLA2III; sPLA2-III | |
| Human recombinant protein fragment corresponding to amino acids 123-238 of human RACGAP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.