missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4D Mouse anti-Human, Clone: 2E2A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66752-1-IG-150UL
This item is not returnable.
View return policy
Description
The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].Specifications
| PLA2G4D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86XP0 | |
| PLA2G4D | |
| PLA2G4D Fusion Protein Ag4178 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2E2A10 | |
| Unconjugated | |
| PLA2G4D | |
| cPLA2 delta, cPLA2delta, Phospholipase A2 group IVD, PLA2G4D | |
| Mouse | |
| Protein G | |
| RUO | |
| 283748 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction