Learn More
Steris™ Plain Top Closure Pouches
Description
- Manufactured from high quality wet strength MG bleached kraft paper
- Double folded bottom seal for extra integrity
- Opening along width
- Conforms to BS 6257
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | BS 6257 |
| Closure Type | Plain |
| Material | Medical Grade Kraft Paper (Bleached) |
| Quantity | 250/Pk. |
| Dimensions (L x W x H) | 300 x 75 x 530mm |
| Length (Metric) | 300 mm |
| Width (Metric) | 75 mm |
| Height (Metric) | 530 mm |