missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasminogen Antibody (C15), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43362
This item is not returnable.
View return policy
Description
Plasminogen Monoclonal antibody specifically detects Plasminogen in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Plasminogen | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen | |
| Immunized with recombinant Human Plasminogen (Uniprot#: P00747; Val98∽Asn810) | |
| 100 μL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Cycle and Replication | |
| 5340 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| C15 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction