missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Plastic Pipette Tips for Graduated Syringe
Description
Together, the HI740143 Graduated Syringe and HI740144P Plastic Pipette Tips are able to accurately transfer 1mL volumes of liquid that need to be precisely dosed. It is ideal for accurate reagent dosing for chemical test kits and photometers.
- 6 piece set
- 1mL volume
- Durable plastic
- Disposable
Specifications
Specifications
| Volume (Metric) | 1 mL |
| Color | Natural |
| For Use With (Application) | Reagent dosing |
| Quantity | 6 pc. |
| Disposable | Single-use |
| For Use With (Equipment) | HI740143 Graduated Syringe |
| Graduated | Yes |
| Material | Plastic |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?