Learn More
Fisherbrand™ Nylon Filter Cloth, 300 mm x 300 mm
Product Code. 11744549 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Pore Size:
5 μm
11 μm
20 μm
30 μm
41μm
53 μm
60 μm
70 μm
100 μm
200 μm
250 μm
315 μm
400 μm
450 μm
500 μm
630 μm
700 μm
800 μm
900 μm
1000 μm
Unit Size:
Pack of 10
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
11744549 41μm Pack of 10
11774569 11 μm Pack of 10
11704569 20 μm Pack of 10
11764539 200 μm Pack of 10
11754539 250 μm Pack of 10
11714539 500 μm Pack of 10
11724549 60 μm Pack of 10
11734549 53 μm Pack of 10
11794539 70 μm Pack of 10
11774539 100 μm Pack of 10
11794529 700 μm Pack of 10
11784569 5 μm Pack of 10
11784529 800 μm Pack of 10
11774529 900 μm Pack of 10
11764529 1000 μm Pack of 10
11754559 30 μm Pack of 10
11744539 315 μm Pack of 10
11734539 400 μm Pack of 10
11724539 450 μm Pack of 10
11704539 630 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 11744549
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Filter Cloths, 300 mm x 300 mm are available in a comprehensive range of pore sizes.
  • Operating temperature of -40° to +115°C
  • Excellent compatibility with alcohols
  • Resistant to aggressive solvents
  • Applications include solvent filtration and particle analysis
Specifications
Pore Size 41μm
Diameter (Metric) Outer 300 mm
Frame Material Plastic
Mesh Material Nylon