Learn More
Platinum Iridium wire, 0.5mm (0.02in) dia, hard
Product Code. 11306767
Change view
Click to view available options
Quantity:
10 cm
50 cm
Unit Size:
10cm
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306767 10 cm 10cm
11316767 50 cm 50cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11306767 Supplier Thermo Scientific Alfa Aesar Supplier No. 010055.BS
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Pt:Ir; 85:15 wt%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Platinum Iridium wire
Quantity 10 cm
Molecular Formula Pt:Ir; 85:15 wt%
MDL Number MFCD02091683
Packaging Shrink-wrapped