Learn More
Platinum powder, amorphous, APS 2.0-8.0 micron, 99.9% (metals basis)
Product Code. 15497405
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497405 1 g 1g
15407415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15497405 Supplier Thermo Scientific Alfa Aesar Supplier No. 047145.03
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 7440-06-4 | Pt | 195.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
Specifications
Quantity 1 g
Odor Odorless
Particle Size APS 2.0 to 8.0 micron
Weight Tap density: 0.5 to 2.5g/cm3
Form Amorphous Powder
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Platinum