missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Library Amplification Master Mix with Primer Mix

Product Code. p-8772009 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
50
250
Unit Size:
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15980523 50 50 reactions
15990523 250 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15980523 Supplier Invitrogen™ Supplier No. A38540050
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen™ Platinum SuperFi Library Amplification Master Mix is a ready-to-use solution designed for amplification of next-generation sequencing (NGS) libraries.

Master mix includes Platinum SuperFi™ DNA Polymerase in combination with a proprietary reaction buffer containing all necessary components for efficient library amplification with minimal bias and superior accuracy.

 Features include:

  • High fidelity—minimal error rates similar to PCR-free methods
  • High efficiency—high yields from minute input amounts
  • Minimal bias—uniform amplification regardless of GC content

 

  • Supplied with a 10X Primer Mix that targets the P5 and P7 regions of Illumina adapters. The master mix is supplemented with an inert blue dye, while the primer mix contains a yellow dye. Mixing both components in a PCR reaction turns the final PCR solution green. This provides a visual aid when pipetting and helps decrease the risk of pipetting errors during reaction setup
  • Platinum SuperFi™ DNA Polymerase has both 5’ to 3’ polymerase and 3’ to 5’ exonuclease (proofreading) activities, but lacks 5’ to 3’ exonuclease activity. Exceptionally strong proofreading activity ensures amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods
  • Proprietary reaction buffer contains all necessary PCR components and additives to enable efficient and uniform library amplification regardless of GC content, helping improve coverage across GC- and AT-rich sequences and other complex regions. High amplification efficiency enables the use of fewer cycles to achieve the desired yield
  • Platinum hot-start technology inhibits DNA polymerase activity at ambient temperatures, allowing room temperature reaction setup and storage of pre-assembled PCR reactions for up to 24 hours prior to the PCR.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 1.25 mL Collibri Library Amplification Master Mix (2X)
• 1 x 500 µL Primer Mix

Store at -20°C.
Sample Type DNA, RNA
Fidelity (vs. Taq) ≥100
GC-Rich PCR Performance High
For Use With (Application) Sequencing
Hot Start Built-In Hot Start
No. of Reactions 50
Overhang Blunt
Product Line Collibri
Product Type Library Amplification Master Mix
Quantity 50 reactions
Reaction Format SuperMix or Master Mix
Shipping Condition Dry Ice
Test Time Standard
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.