missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121679
This item is not returnable.
View return policy
Description
PLB1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
This gene encodes a membrane-associated phospholipase that displays lysophospholipase and phospholipase A2 activities through removal of sn-1 and sn-2 fatty acids of glycerophospholipids. In addition, it displays lipase and retinyl ester hydrolase activities. The encoded protein is highly conserved and is composed of a large, glycosylated extracellular domain composed of four tandem homologous domains, followed by a hydrophobic segment that anchors the enzyme to the membrane and a short C-terminal cytoplasmic tail. This gene has been identified as a candidate rheumatoid arthritis risk gene.
Specifications
| PLB1 | |
| Polyclonal | |
| Unconjugated | |
| PLB1 | |
| 4632413E21Rik; 4930433E17Rik; 4930539A06Rik; BC033606; hPLB; Lysophospholipase; Phlpb; Phospholipase A2; phospholipase B; Phospholipase B/lipase; phospholipase B1; phospholipase B1, membrane-associated; PLB; PLB/LIP; PLB1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 151056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6P1J6 | |
| PLB1 | |
| Recombinant protein Phospholipase B. The antigen corresponds to amino acid range 1036-1458 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction