missing translation for 'onlineSavingsMsg'
Learn More

PLCB3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51139-1-AP-150UL

 View more versions of this product

Product Code. 16826405

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

PLCB3
Polyclonal
Unconjugated
PLCB3
Phospholipase C beta 3, PLC beta 3, PLCB3
Rabbit
Antigen Affinity Chromatography
RUO
5331
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q01970
PLCB3
Peptide
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.