missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19143-1-AP-150UL
This item is not returnable.
View return policy
Description
PLCH1 is a member of the PLC-eta family of the phosphoinositide-specific phospholipase C (PLC) superfamily of enzymes that cleave phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to generate second messengers inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG).Specifications
| PLCH1 | |
| Polyclonal | |
| Unconjugated | |
| PLCH1 | |
| KIAA1069, Phospholipase C eta 1, Phospholipase C like protein 3, phospholipase C, eta 1, PLC eta 1, PLC L3, PLCeta1, PLCH1, PLCL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23007, 269437, 310463 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4KWH5, Q4KWH8 | |
| PLCH1 | |
| PLCH1 Fusion Protein Ag13607 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction