Learn More
PLCL2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17421-1-AP-150UL
Description
The PLCL2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PLCL2. This antibody recognizes human, mouse, rat antigen. The PLCL2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PLCL2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 23228 | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |