missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PLD3 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807204
|
Proteintech
17327-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897194
|
Proteintech
17327-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.Specifications
| PLD3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O35405, Q5FVH2, Q8IV08 | |
| 18807, 23646, 361527 | |
| PLD3 Fusion Protein Ag11306 | |
| Primary | |
| -20°C | |
| PLD3 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Choline phosphatase 3, HindIII K4L homolog, HU K4, Phospholipase D3, PLD 3, PLD3 | |
| Pld3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title