Learn More
Plexin B2 Antibody (3E7) - BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43310-0.025MG
This item is not returnable.
View return policy
Description
Plexin B2 Monoclonal specifically detects Plexin B2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Plexin B2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2 | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
| Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunohistochemistry | |
| 30000000 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| PLXNB2 | |
| The immunogen for this antibody was PlexinB2. | |
| 0.025 mg | |
| Growth and Development, Neuronal Cell Markers | |
| 23654 | |
| Store at 4C. Do not freeze. |