Learn More
PLG Monoclonal Antibody (OTI4F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810638
Description
Plasmin (PLG, Plasminogen) dissolves the fibrin of blood clots and acts as a proteolytic factor in a variety of other processes including embryonic development, tissue remodeling, tumor invasion, and inflammation. In ovulation, weakens the walls of the Graafian follicle. It activates the urokinase-type plasminogen activator, collagenases and several complement zymogens, such as C1 and C5. Cleavage of fibronectin and laminin leads to cell detachment and apoptosis. Also cleaves fibrin, thrombospondin and von Willebrand factor. Its role in tissue remodeling and tumor invasion may be modulated by CSPG4. Plasmin binds to cells.Specifications
| PLG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00747 | |
| PLG | |
| Human recombit protein fragment corresponding to amino acids 301-586 of human PLG produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI4F5 | |
| Unconjugated | |
| PLG | |
| plasminogen | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5340 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.