missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21299-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects both PLK5 gene of human and mouse.
Inactive serine/threonine-protein kinase that plays a role in cell cycle progression and neuronal differentiation. Ota T., Nat. Genet. 36:40-45(2004). Li H., Submitted (MAR-2006) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004).Specifications
| PLK5 | |
| Polyclonal | |
| Unconjugated | |
| PLK5 | |
| PLK5, PLK5P, polo like kinase 5 pseudogene, SgK384ps | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 126520, 216166 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q496M5, Q4FZD7 | |
| Plk5 | |
| PLK5 Fusion Protein Ag15881 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction