Learn More
SI Analytics™ Connection Cables and Plugs
Product Code. 10684661 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric) Cable:
1 m
2 m
5 m
10 m
Type:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 1 R
L 2 A
Plug Cable Combination
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Type Unit Size
10684661 1 m L 1 A Each
10001072 1 m L 1 NN Each
10535553 5 m K 10 A Each
10326932 1 m L 1 N Each
10604671 1 m L 1 BNC Each
10406812 1 m K 1 A Each
10305111 1 m L 1 R Each
10466165 2 m L 2 A Each
10652832 5 m Plug Cable Combination Each
10152071 2 m K2X Each
10559371 10 m K5A Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10684661 Supplier SI Analytics™ Supplier No. 285122456
  • ONLINE EXCLUSIVE PRICE
    70.00€
    67.00€ / Pack of 1
    Save 3.00€
    4% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Connector Types DIN
For Use With (Equipment) Connector Electrodes for pH and ORP Electrodes with Plug Head
Type L 1 A
Includes 1m Coaxial Cable, DIN Instrument Plug
Length (Metric) Cable 1 m