Learn More
Invitrogen™ PMEL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583201
Description
Immunogen sequence: PTAESTGMTP EKVPVSEVMG TTLAEMSTPE ATGMTPAEVS IVVLSGTTAA QVTTTEWVET TARELPIPEP EGPDASSIMS TESITGSLGP LLDGTATLRL VKRQVPLDCV LYRYGSFSVT LDIVQGIESA EILQAV Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 67%.
Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.
Specifications
| PMEL | |
| Polyclonal | |
| Unconjugated | |
| PMEL | |
| 95 kDa melanocyte-specific secreted glycoprotein; 95kDa melanocyte-specific secreted glycoprotein; D10H12S53E; D12S53E; D12S53Eh; gp100; gp87; M-alpha; M-beta; ME20; ME20M; ME20-M; ME20-S; Melanocyte lineage specific antigen GP100; melanocyte protein mel 17; melanocyte protein PMEL; Melanocyte protein Pmel 17; melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; P26; PMEL; Pmel 17; PMEL17; premelanosome protein; Secreted melanoma-associated ME20 antigen; SI; SIL; SILV; silver homolog; silver locus protein; silver locus protein homolog; silver, mouse, homolog of | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6490 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P40967 | |
| PMEL | |
| Recombinant protein corresponding to Human PMEL. Recombinant protein control fragment (Product #RP-102598). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.