Learn More
Kartell SpA PMP Graduated Beaker
Product Code. 10001121
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
3000 mL
Unit Size:
Pack of 2
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10001121 3000 mL Pack of 2
10122281 25 mL Pack of 20
10325731 50 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10001121 Supplier Kartell SpA Supplier No. 0133000
  • ONLINE EXCLUSIVE PRICE
    53.00€
    49.95€ / Pack of 2
    Save 3.05€
    5% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Graduated beakers, low form - Class B
Conforms to ISO 7056 - 1981 (E) . Crystal clear, autoclavable. Will withstand 170°C for short periods. Permanent graduations are moulded in for reproducibility. Excellent chemical resistance.
Specifications
Capacity (Metric) 3000 mL
Material PMP
Type Graduated Beaker
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Molded Scale
Style Griffin, Low Form