missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS1 Antibody (CL14436) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26854G
This item is not returnable.
View return policy
Description
PMS1 Monoclonal antibody specifically detects PMS1 in Human samples. It is validated for Western Blot
Specifications
| PMS1 | |
| Monoclonal | |
| DyLight 488 | |
| DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E | |
| This antibody was developed using a synthetic peptide derived from P54277, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers | |
| 5378 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot | |
| CL14436 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction