Learn More
PMVK Monoclonal Antibody (OTI1C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503447S
Description
PMVK Monoclonal antibody specifically detects PMVK in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| PMVK | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q07343 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5142 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI1C12 | |
| Unconjugated | |
| PDE4B | |
| 1110011E12Rik; 2900002L22Rik; AV001327; hPMK; HUMPMKI; Phosphomevalonate kinase; PMK; PMKA; PMKASE; PMKI; Pmvk; POROK1; testis tissue sperm-binding protein Li 95mP | |
| Full length human recombinant protein of human PDE4B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.