missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNKP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13558-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene phosphorylates vitamin B6, a step required for the conversion of vitamin B6 to pyridoxal-5-phosphate, an important cofactor in intermediary metabolism. The encoded protein is cytoplasmic and probably acts as a homodimer. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| PNKP | |
| Polyclonal | |
| Unconjugated | |
| PNKP | |
| DNA 5 kinase/3 phosphatase, PNK, PNKP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11284, 308576, 59047 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96T60, Q9JLV6 | |
| Pnkp | |
| PNKP Fusion Protein Ag4421 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction