missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PNP Monoclonal Antibody (773511)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561671

Product Code. 30534501

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The PNP gene encodes for the enzyme purine nucleoside phosphorylase, which plays a vital role in the purine salvage pathway by catalyzing the reversible cleavage of purine nucleosides. This enzyme facilitates the conversion of inosine, guanosine, and deoxyguanosine into free bases and sugars phosphorylated at the C1 position. PNP is located on chromosome 14q13 and is crucial for nucleoside catabolism, impacting energy metabolism and cellular functions across various organisms. PNP deficiency is an autosomal recessive multisystem disorder arising from mutations in the PNP gene, leading to impaired T-cell maturation and function, resulting in recurrent infections, autoimmune manifestations, and neurological abnormalities due to the toxic accumulation of deoxyguanosine triphosphate (dGTP). This deficiency particularly affects immature thymocytes and can lead to lymphoid and neuronal toxicity, underscoring the gene's importance in proper immune system function and cellular health.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PNP
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P00491
PNP
E. coli-derived recombinant human Purine Nucleoside Phosphorylase/PNP, Met1-Ser289.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
773511
Unconjugated
PNP
AL024301; AU015798; endothelial dysfunction inducing protein; epididymis secretory sperm binding protein Li 156an; HEL-S-156an; inosine phosphorylase; inosine phosphorylase {ECO:0000250; Inosine-guanosine phosphorylase; Np; Np-1; Np-2; nucleoside phosphorylase; PNP; PNP {ECO:0000250; Pnp1; PRO1837; PUNP; purine nucleoside phosphorylase; purine nucleoside phosphorylase {ECO:0000250; purine-nucleoside phosphorylase; purine-nucleoside phosphorylase 1; purine-nucleoside:orthophosphate ribosyltransferase; UniProtKB:P00491}
Mouse
Protein A/G
RUO
4860
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.