missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PNPLA8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524642

Product Code. 13211849

  • 518.00€ / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PNPLA8 Polyclonal Antibody for Western Blot, Flow

Phospholipase A2 catalyzes cleavage of fatty acids from phospholipids, thereby regulating membrane physical properties and the release of lipid second messengers and growth factors. Phospholipase A2 activity also modulates cellular growth programs, inflammation, and ion channel function (Mancuso et al., 2000 [PubMed 10744668]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PNPLA8
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q9NP80
PNPLA8
KLH conjugated synthetic peptide between 212-240 amino acids from the N-terminal region of human PNPLA8.
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot
Unconjugated
PNPLA8
1200006O19Rik; AI467579; BM-043; Calcium-independent phospholipase A2-gamma; intracellular membrane-associated calcium-independent phospholipase A2 gamma; iPLA2 gamma; Ipla2(gamma); IPLA22; IPLA2-2; IPLA2G; iPLA2gamma; iPLA2-gamma; membrane-associated calcium-independent phospholipase A2 gamma; MMLA; patatin like phospholipase domain containing 8; patatin-like phospholipase domain containing 8; patatin-like phospholipase domain-containing protein 8; Pnpla8; PNPLA-gamma
Rabbit
Antigen affinity chromatography, Protein A
RUO
50640
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.