missing translation for 'onlineSavingsMsg'
Learn More

PNU 142300, TRC

Product Code. 17984735
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
0.25mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984735 0.25 mg 0.25mg
17972955 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984735 Supplier Toronto Research Chemicals Supplier No. TRCP6534000.25MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368891-69-4
Molecular Formula C16H20FN3O6
Molecular Weight (g/mol) 369.34
Synonym N-[4-[(5S)-5-[(Acetylamino)methyl]-2-oxo-3-oxazolidinyl]-2-fluorophenyl]-N-(2-hydroxyethyl)glycine;
SMILES CC(=O)NC[C@H]1CN(C(=O)O1)C2=CC(=C(C=C2)NCCOCC(=O)O)F

Specifications

Quantity 0.25 mg
InChI Formula InChI=1S/C16H20FN3O6/c1-10(21)19-7-12-8-20(16(24)26-12)11-2-3-14(13(17)6-11)18-4-5-25-9-15(22)23/h2-3,6,12,18H,4-5,7-9H2,1H3,(H,19,21)(H,22,23)/t12-/m0/s1
Formula Weight 369.1336
Percent Purity >95%
Chemical Name or Material PNU 142300
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.