missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin Like Antibody (CL0308) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52912F
This item is not returnable.
View return policy
Description
Podocalyxin Like Monoclonal antibody specifically detects Podocalyxin Like in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Podocalyxin Like | |
| Monoclonal | |
| FITC | |
| Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
| This antibody was developed using a recombinant protein derived from O00592, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 5420 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0308 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction