missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Podocin Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543645
This item is not returnable.
View return policy
Description
The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.
Specifications
| Podocin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8K4G9, Q91X05, Q9NP85 | |
| Nphs2 | |
| Met222-Pro372 (Accession Q9NP85), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Rat | |
| Antibody | |
| IgG2a κ |
| Western Blot | |
| C3 | |
| Unconjugated | |
| Nphs2 | |
| AI790225; nephrosis 2 homolog, podocin; nephrosis 2 idiopathic steroid-resistant (podocin); nephrosis 2, idiopathic, steroid-resistant; nephrosis 2, idiopathic, steroid-resistant (podocin); nephrosis 2, podocin; NPHS2; NPHS2 podocin; NPHS2 stomatin family member, podocin; NPHS2, podocin; PDCN; podocin; SRN1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 170484, 170672, 7827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction