missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (PDPN/1433) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54347-100UG
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal specifically detects Podoplanin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.
Specifications
| Podoplanin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Mouse | |
| 41 kDa | |
| 100 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| PDPN/1433 | |
| Unconjugated | |
| PBS with No Preservative | |
| PDPN | |
| Recombinant human Podoplanin (PDPN) protein fragment (aa24-126) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction