Learn More
Thermosafe™ PolarPack™ Refrigerant Gel Packs
Product Code. 12201372
Change view
Click to view available options
Weight (English):
8 oz./Pack
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
12201372 8 oz./Pack Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12201372 Supplier Thermosafe™ Supplier No. THV245020P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Economical and practical refrigerant pack choice for shipping temperature-sensitive material such as food products, pharmaceuticals, and more. Thermosafe™ PolarPack™ Refrigerant Gel Packs are enclosed in durable polyethylene film.
  • Reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products.
  • Multi-layered exterior minimizes surface condensation which protects product from moisture damage.
  • Outer film layer assures products arrive in top condition by absorbing condensation.
  • Phase change formulas ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2.
  • Laminated, metalized film exterior provides radiant heat barrier, resists punctures.
  • Available in several sizes
  • Reusable
  • Non-toxic
TRUSTED_SUSTAINABILITY
Specifications
Quantity 72 Pk./Cs.
Weight (English) 8 oz./Pack
Dimensions (LxWxH) 6.5 x 3.5 x 0.75 in.