Learn More
POLI Monoclonal Antibody (OTI5H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801479S
Description
POLI Monoclonal antibody specifically detects POLI in Human samples. It is validated for Western BlotSpecifications
| POLI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UNA4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11201 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5H9 | |
| Unconjugated | |
| Poli | |
| DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB | |
| Human recombinant protein fragment corresponding to amino acids 458-740 of human POLI produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.