missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLL Recombinant Rabbit Monoclonal Antibody (PSH05-24)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA556437

Product Code. 30275863

  • 500.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POLL Recombinant Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLL
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q9QXE2, Q9UGP5
Poll
Recombinant protein within human DNA Polymerase lambda aa 1-575/575.
100 μL
Primary
Human, Mouse, Monkey
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
PSH05-24
Unconjugated
Poll
1110003P06Rik; AV007317; BETAN; DNA polymerase beta-2; DNA polymerase beta-N; DNA polymerase kappa; DNA polymerase lambda; DNA-directed DNA polymerase lambda; EC 2.7.7.7; FLJ46002; OTTHUMP00000020323; Pol beta2; Pol Lambda; Polk; POLKAPPA; POL-KAPPA; Poll; polymerase (DNA directed), lambda; polymerase (DNA) lambda; RP11-529I10.1
Rabbit
Protein A
RUO
0, 27343, 56626
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.