missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13428-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the third largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains a cysteine rich region and exists as a heterodimer with another polymerase subunit, POLR2J. These two subunits form a core subassembly unit of the polymerase. A pseudogene has been identified on chromosome 21.Specifications
| POLR2C | |
| Polyclonal | |
| Unconjugated | |
| POLR2C | |
| hRPB33, hsRPB3, POLR2C, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20021, 361365, 5432 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19387, P97760 | |
| Polr2c | |
| POLR2C Fusion Protein Ag4228 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction