Learn More
Invitrogen™ POLR2C Recombinant Rabbit Monoclonal Antibody (24GB1535)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557235
This item is not returnable.
View return policy
Description
POLR2C Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft.
Specifications
| POLR2C | |
| Recombinant Monoclonal | |
| Unconjugated | |
| POLR2C | |
| 33kDa; A-152E5.7; DNA-directed RNA polymerase II 33 kDa polypeptide; DNA-directed RNA polymerase II subunit C; DNA-directed RNA polymerase II subunit RPB3; hRPB33; hsRPB3; mRBP31; Polr2c; polymerase (RNA) II (DNA directed) polypeptide C; polymerase (RNA) II (DNA directed) polypeptide C, 33kDa; polymerase (RNA) II subunit C; polymerase II polypeptide C; RNA polymerase II 3; RNA polymerase II subunit 3; RNA polymerase II subunit B3; RNA polymerase II subunit C; RPB3; RPB31; RPB33; Rpo2-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20021, 361365, 5432 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1535 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19387, P97760 | |
| POLR2C | |
| A synthesized peptide derived from human POLR2C (aa 50-100). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |