Learn More
POLR2F Antibody (OTI2F2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44902-30ul
 View more versions of this product
Product Code. 30306308
  • 256.00€ / 30µL
Description
POLR2F Monoclonal antibody specifically detects POLR2F in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POLR2F
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot
OTI2F2
Western Blot 1:2000
DNA-directed RNA polymerase II subunit F, DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide, HRBP14.4, II, and III subunit RPABC2, polymerase (RNA) II (DNA directed) polypeptide F, RNA Polymerase II subunit 14.4 kD, RNA polymerases I, II, and III subunit ABC2, RPB6, RPB6 homolog
Full length human recombiant protein of human POLR2F (NP_068809) produced in E.coli.
30 μL
Core ESC Like Genes, Stem Cell Markers
5435
Store at -20 degrees C. Avoid freeze/thaw cycles.