missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific™

132.00€ - 516.00€

Chemical Identifiers

CAS 104934-50-1
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00217686
InChI Key DUFPJSOXRHVDOV-UHFFFAOYSA-N
Synonym poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
PubChem CID 22950249
IUPAC Name 3-hexyl-2,5-dimethylthiophene
SMILES CCCCCCC1=C(SC(=C1)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15436615
SDS View Documents
Thermo Scientific Alfa Aesar
044745.77
0.1g
132.00€
0.1g
View alternative products
Save Up to 
»
Estimated Shipment: 14-09-2026
Add to basket
 
 Restricted Item
15426615
SDS View Documents
Thermo Scientific Alfa Aesar
044745.02
0.5g
516.00€
0.5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

104934-50-1
196.352
DUFPJSOXRHVDOV-UHFFFAOYSA-N
22950249
CCCCCCC1=C(SC(=C1)C)C
C12H20S
MFCD00217686
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
3-hexyl-2,5-dimethylthiophene

Specifications

104934-50-1
MFCD00217686
Soluble in Chloroform and tetrahydrofuran
CCCCCCC1=C(SC(=C1)C)C
196.352
Poly (3-hexylthiophene-2,5-diyl), Regioregular, low metals
C12H20S
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
DUFPJSOXRHVDOV-UHFFFAOYSA-N
3-hexyl-2,5-dimethylthiophene
22950249
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.