missing translation for 'onlineSavingsMsg'
Learn More

Poly(allylamine Hydrochloride) (Technical Grade), TRC

Product Code. 30412850
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412850 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412850 Supplier Toronto Research Chemicals Supplier No. TRCP6883032.5G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyallylamine Hydrochloride (2-propen-1-amine hydrochloride','homopolymer)
CAS 71550-12-4
Molecular Formula (C3 H8 Cl N)mon
Quantity 2.5 g
Synonym Allylamine hydrochloride homopolymer, Allylamine hydrochloride polymer, Allylammonium chloride homopolymer, Danfix 723, Monorex RD
InChI Formula InChI=1S/C3H7N.ClH/c1-2-3-4;/h2H,1,3-4H2;1H
SMILES NCC=C.Cl
Molecular Weight (g/mol) 93.56
Formula Weight 93.0345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.