missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol) Methyl Ether Mn~2000, TRC

Product Code. 30179073
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179073 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179073 Supplier Toronto Research Chemicals Supplier No. TRCP6885235G
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-74-4
Molecular Formula CH3O(CH2CH2O)nH
Quantity 5 g
Synonym (Methoxypolyethylene Glycol), α-Methyl-ω-hydroxypoly(oxy-1,2-ethanediyl), Polyoxyethylene Monomethyl Ether, Poly(Ethylene Oxide) Methyl Ether, Poly(Ethylene Oxide) Monomethyl Ether;
SMILES OCCOC
Chemical Name or Material Methoxypolyethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.