missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol) Methyl Ether Mn~5000, TRC

Product Code. 30179540
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179540 100 g 100g
30179399 50 g 50g
30179079 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179540 Supplier Toronto Research Chemicals Supplier No. TRCP688525100G
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-74-4
Molecular Formula CH3O(CH2CH2O)nH
Synonym (Methoxypolyethylene Glycol), α-Methyl-ω-hydroxypoly(oxy-1,2-ethanediyl), Polyoxyethylene Monomethyl Ether, Poly(Ethylene Oxide) Methyl Ether, Poly(Ethylene Oxide) Monomethyl Ether;
SMILES OCCOC

Specifications

Quantity 100 g
Chemical Name or Material Methoxypolyethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.