Learn More
MP Biomedicals™ Poly-L-Lysine Hydrochloride
Product Code. 17348583 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17348583 500 mg 500mg
17338583 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17348583 Supplier MP Biomedicals™ Supplier No. 0215269090
  • ONLINE EXCLUSIVE PRICE
    1905.00€
    1698.56€ / 500mg
    Save 206.44€
    10% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Poly-L-Lysine Hydrochloride is a cellular attachment factor in culture applications.
  • Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
  • Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
  • When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
  • Polymers of both D- and L-lysine are used to coat solid surfaces.
  • Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
Specifications
CAS 26124-78-7
Color Whitish
Content And Storage -20°C
Format Powder
For Use With (Application) Cellular attachment factor in culture applications
Molecular Weight (g/mol) >20,000 g/mol
Quantity 500 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.