missing translation for 'onlineSavingsMsg'
Learn More

Poly-Ubiquitin Antibody (HWA4C4) - K63 Linkage, Novus Biologicals™

Mouse Monoclonal Antibody

276.00€ - 430.00€

Specifications

Antigen Poly-Ubiquitin
Clone HWA4C4
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18143579
View Documents
Novus Biologicals
NBP2-36523-25UL
25 μL
276.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18038893
View Documents
Novus Biologicals
NBP2-36523
0.05 mL
430.00€
0.05mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Poly-Ubiquitin
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
Unconjugated
RUO
PBS with 0.09% Sodium Azide
6233
IgG2a
Ion exchange chromatography
HWA4C4
Monoclonal
Mouse
Non-species specific
CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80
The hybridoma secreting the antibody was generated by fusion of splenocytes from Balb/c mice which had received repeated immunisation with a synthetic peptide encompassing the ubiquitin K63-isopeptide motif.
Primary
Antibody specifically recognizes K63-linked polyubiquitin chains, but not any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitin chain or monoubiquitin. The specificity of this K63-linked ubiquitin specific monoclonal antibody was demonstrated using synthetic ubiquitin chains, single isopeptide-linked polyubiquitin chains, and with endogenously ubiquitinylated species in both cell lysates and intact cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.