missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl Cinnamate) ( average Mn 40,000, powder), TRC

Product Code. 30426521
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30426521 2.5 g
30416978 250 mg
30410537 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426521 Supplier Toronto Research Chemicals Supplier No. TRCP6991702.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9050-06-0
Molecular Formula (C11H10O2)nC2H6
IUPAC Name butan-2-yl (E)-3-phenylprop-2-enoate
SMILES CCC(C)OC(=O)\C=C\c1ccccc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H16O2/c1-3-11(2)15-13(14)10-9-12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/b10-9+
Formula Weight 204.115
Chemical Name or Material Poly(vinyl Cinnamate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.