missing translation for 'onlineSavingsMsg'
Learn More

Polyanetholesulfonic Acid Sodium Salt, Reagent Grade, MP Biomedicals™

Product Code. 11426322 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426322 5 g 5g
11436322 25 g 25g
11446322 100 g 100g
11456322 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11426322 Supplier MP Biomedicals Supplier No. 0210292305
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 52993-95-0
Molecular Formula (C10H11O4SNa)n
MDL Number MFCD00148427
Synonym sodium anethole, benzenesulfonic acid, methoxypropenyl-, sodium salt, homopolymer, sodium methoxypropenylbenzenesulonate homopolymer, benzenesulfonic acid, methoxy 1-propenyl-, sodium salt, homopolymer, sodium 1-methoxy-4-e-prop-1-enyl benzene, benzenesulfonic acid, methoxy 1-propen-1-yl-, sodium salt 1:1 , homopolymer

Specifications

Color White
Quantity 5 g
Formula Weight (250)n
Grade Reagent
Physical Form Solid
Chemical Name or Material Polyanetholesulfonic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.