Learn More
Masterflex™ Polycarbonate Female Luer Fitting
Product Code. 15819911 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
Fits Female luer x 1/4 in. hose barb adapter
Fits Female luer x 3/16 in. hose barb adapter
Fits Female luer x 4mm (5/32 in.) hose barb adapter
Fits Female luer x female luer adapter
Fits Female x Female x Female
Unit Size:
Each
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
15819911 Fits Female luer x 4mm (5/32 in.) hose barb adapter Pack of 25
13538810 Fits Female luer x 3/16 in. hose barb adapter Pack of 25
13477728 Fits Female luer x female luer adapter Pack of 25
13551060 Fits Female luer x 4mm (5/32 in.) hose barb adapter Pack of 25
13518810 Fits Female luer x 1/4 in. hose barb adapter Pack of 25
13487728 Fits Female x Female x Female Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15819911 Supplier Masterflex™ Supplier No. 4550100
  • ONLINE EXCLUSIVE PRICE
    27.05€
    22.81€ / Pack of 25
    Save 4.24€
    15% Off
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Material Polycarbonate
For Use With Fits Female luer x 4mm (5/32 in.) hose barb adapter
Packaging Quantity 25 Pack
Product Type Female Luer Fitting