Learn More
Brady™ Polyester Labels
Product Code. 13533320 Shop All Brady Products
Change view
Click to view available options
Adhesive Type:
Acrylic, Permanent
Permanent Rubber
Width (Metric):
25.4 mm
48.26 mm
50.8 mm
Height (Metric):
6.35 mm
12.70 mm
25.40 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Adhesive Type Width (Metric) Height (Metric) Unit Size
13533320 Permanent Rubber 25.4 mm 12.70 mm Each
13523320 Acrylic, Permanent 50.8 mm 25.40 mm Each
13563320 Acrylic, Permanent 48.26 mm 6.35 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13533320 Supplier Brady™ Supplier No. 143075
  • ONLINE EXCLUSIVE PRICE
    146.00€
    127.08€ / Pack of 1
    Save 18.92€
    12% Off
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Polyester Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant.
Specifications
Certifications/Compliance ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognised
Description Polyester Labels for BBP33/i3300 Printers
Adhesive Type Permanent Rubber
Coating Powder
Color White
Display Blank
Finish Gloss
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset tracking, Barcode labelling, Component and equipment labelling, Data and telecommunications labelling, Electrical labelling, General Identification, Inventory and inspection labelling
For Use With (Equipment) BBP33, i3300, i5300
Horizontal Repeat (Metric) 27.3 mm
Labels per Row 3
Min. Operating Temperature (Metric) -40°C
No. per Pack 5000 Labels/Box
Quantity 5000 Pk.
Shape Rectangle
Software Type LabelMark, MarkWare
Temperature Range (English) -40°F to +248°F
Temperature Range (Metric) -40°C to +120°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 15.24 mm
Dimensions (L x W) 12.7 x 25.4 mm
Length (English) 0.5 in.
Length (Metric) 12.7 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (Metric) 12.70 mm
Alternate Part Number B33-5-483