missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Polyethylene Coveralls

Product Code. 11866920
Change view
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11866920 X-Large Pack of 25
11856910 Medium Pack of 25
11866910 Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11866920 Supplier Sperian™ Supplier No. 4180161XL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Single-use Coveralls
Size X-Large
Gender Unisex
Material Polyethylene
Color Green
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Sterility Non-sterile
Protection Class Class 3
Resistance Abrasion, Tears, Perforation
Certifications/Compliance (Europe) EC Category PPE, Cat III to Directive 89/686/EEC
Thickness 150
For Use With (Application) Chemical Industries, Maintenance, Medical and Pharmaceuticals, Spray Painting, Automotive
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.